MilliporeSigma
  • Core-shell column Tanaka characterization and additional tests using active pharmaceutical ingredients.
Core-shell column Tanaka characterization and additional tests using active pharmaceutical ingredients.
Journal of separation science (2016-10-16)
Jufang Wu Ludvigsson, Anders Karlsson, Viktor Kjellberg
ABSTRACT
In the last decade, core-shell particles have gained more and more attention in fast liquid chromatography separations due to their comparable performance with fully porous sub-2 μm particles and their significantly lower back pressure. Core-shell particles are made of a solid core surrounded by a shell of classic fully porous material. To embrace the developed core-shell column market and use these columns in pharmaceutical analytical applications, 17 core-shell C
Sigma-Aldrich
o-Terphenyl, 99%
Pricing and availability is not currently available.