Merck
p21-activated kinases: three more join the Pak.
The international journal of biochemistry & cell biology (2002-04-16)
Zahara M Jaffer, Jonathan Chernoff
ABSTRACT
The p21-activated kinases (Paks) are serine/threonine protein kinases that bind to and, in some cases, are stimulated by activated forms of the small GTPases, Cdc42 and Rac. With the recent discovery of several novel isoforms, Paks are now categorized into two subgroups based on architectural similarities. The Group I Paks (Pak1, Pak2, Pak3) have been studied in greater detail and shown to be involved in the regulation of cellular processes such as gene transcription, cell morphology, motility, and apoptosis. Here we summarize recent findings that shed light on the newly recognized Group II Paks (Pak4, Pak5, Pak6) and review both similarities and differences between kinases of the two Pak subgroups.
Sigma-Aldrich
PAK3 Protein, active, 10 µg, Active, C-terminal 6His-tagged, recombinant, human full length, PAK3. For use in Kinase Assays.
Sigma-Aldrich
PAK3, active, GST tagged from mouse, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
PAK7, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
PAK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution