Merck
  • NADPH oxidase 4 is involved in the triethylene glycol dimethacrylate-induced reactive oxygen species and apoptosis in human embryonic palatal mesenchymal and dental pulp cells.
NADPH oxidase 4 is involved in the triethylene glycol dimethacrylate-induced reactive oxygen species and apoptosis in human embryonic palatal mesenchymal and dental pulp cells.
Clinical oral investigations (2014-12-04)
Cheng-Chang Yeh, Jenny Zwei-Chieng Chang, Wan-Hsien Yang, Hao-Hueng Chang, Eddie Hsiang-Hua Lai, Mark Yen-Ping Kuo
ABSTRACT
Triethylene glycol dimethacrylate (TEGDMA) is a common component of resin-based dental composites and endodontic sealers. TEGDMA induces apoptosis in several types of cells. However, the mechanisms are not completely understood. The aim of this study was to investigate the mechanisms underlying TEGDMA-induced apoptosis in human embryonic palatal mesenchymal (HEPM) pre-osteoblasts and primary human dental pulp (HDP) cells. Cell viability was examined after TEGDMA treatment. Cell cycle progression was checked by flow cytometry. Apoptotic cells were evaluated using terminal deoxynucleotidyl transferase (TdT)-mediated dUTP nick end labeling assay and visualized by fluorescence microscopy. Western blot analyses were performed to determine expressions of apoptosis-related proteins. The production of reactive oxygen species (ROS) was detected using flow cytometry. NADPH oxidase 4 (NOX4) expression levels were investigated using real-time quantitative polymerase chain reaction and Western blot analyses. TEGDMA increased cytosol cytochrome c levels and activated caspase-9 in HEPM and HDP cells. TEGDMA decreased the expression of anti-apoptotic protein Bcl-XL. TEGDMA-induced apoptosis was inhibited by caspase-9-specific inhibitor, anti-oxidants, NOX inhibitor, NOX4 inhibitor, and NOX4 small interfering RNA (siRNA). TEGDMA increased ROS production and upregulated NOX4 mRNA and protein expression. TEGDMA-induced intracellular ROS production was inhibited by NOX inhibitor and NOX4 inhibitor. We demonstrate significant involvement of NOX4 in the TEGDMA-induced ROS. NOX4-derived ROS subsequently induces mitochondrial cytochrome c release leading to apoptosis through activation of the intrinsic apoptotic pathway. NOX4 may be a potential target for strategies to prevent or ameliorate the TEGDMA-induced toxicity in HEPM and HDP cells.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nox4
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
MISSION® esiRNA, targeting human NOX4
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Plumbagin from Plumbago indica
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%