Merck
  • Preparation of a polyclonal antibody that recognizes a unique galactoseβ1-4fucose disaccharide epitope.
Preparation of a polyclonal antibody that recognizes a unique galactoseβ1-4fucose disaccharide epitope.
Carbohydrate research (2015-05-31)
Tomoharu Takeuchi, Kazusa Nishiyama, Saori Saito, Mayumi Tamura, Takashi J Fuwa, Shoko Nishihara, Hideyo Takahashi, Hideaki Natsugari, Yoichiro Arata, Ken-ichi Kasai
ABSTRACT
Galactoseβ1-4fucose (Galβ1-4Fuc) is a unique disaccharide unit that has been found only in the N-glycans of protostomia. We demonstrated that this unit has a role as an endogenous ligand for Caenorhabditis elegans galectins. This unit is also recognized by fungal and mammalian galectins possibly as a non-self glycomarker. In order to clarify its biological function, we made a polyclonal antibody using (Galβ1-4Fuc)n-BSA as the antigen, which was prepared by crosslinking Galβ1-4Fuc-O-(CH2)2-SH and BSA. The binding specificity of the antibody was analyzed by frontal affinity chromatography, and it was confirmed that it recognizes naturally occurring N-glycans containing the Galβ1-4Fuc unit linked to the reducing-end GlcNAc via α1-6 linkage. By western blotting analysis, the antibody was also found to bind to (Galβ1-4Fuc)n-BSA but not to BSA or asialofetuin, which has N-glycan chains containing Galβ1-4GlcNAc. Western blotting experiments also revealed presence of stained proteins in crude extracts of C. elegans, the parasitic nematode Ascaris suum, and the allergenic mite Dermatophagoides pteronyssinus, while those from Drosophila melanogaster, Mus musculus, and the allergenic mites Dermatophagoides farinae and Tyrophagus putrescentiae were negative. This antibody should be a very useful tool for research on the distribution of the Galβ1-4Fuc disaccharide unit in glycans in a wide range of organisms.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%