Merck
  • tert-Butyl peroxybenzoate (TBPB)-mediated 2-isocyanobiaryl insertion with 1,4-dioxane: efficient synthesis of 6-alkyl phenanthridines via C(sp3)-H/C(sp2)-H bond functionalization.
tert-Butyl peroxybenzoate (TBPB)-mediated 2-isocyanobiaryl insertion with 1,4-dioxane: efficient synthesis of 6-alkyl phenanthridines via C(sp3)-H/C(sp2)-H bond functionalization.
Chemical communications (Cambridge, England) (2014-04-05)
Jia-Jia Cao, Tong-Hao Zhu, Shun-Yi Wang, Zheng-Yang Gu, Xiang Wang, Shun-Jun Ji
ABSTRACT
An efficient method for the construction of 6-alkyl phenanthridines by tert-butyl peroxybenzoate (TBPB)-mediated 2-isocyanobiaryl insertion with 1,4-dioxane was established. Two new C-C bonds were formed in this reaction via a sequential C(sp(3))-H/C(sp(2))-H bond functionalization under metal-free conditions.
Sigma-Aldrich
Luperox® P, tert-Butyl peroxybenzoate, 98%