Merck
  • Genetic signatures of a demographic collapse in a large-bodied forest dwelling primate (Mandrillus leucophaeus).
Genetic signatures of a demographic collapse in a large-bodied forest dwelling primate (Mandrillus leucophaeus).
Ecology and evolution (2012-07-24)
Nelson Ting, Christos Astaras, Gail Hearn, Shaya Honarvar, Joel Corush, Andrew S Burrell, Naomi Phillips, Bethan J Morgan, Elizabeth L Gadsby, Ryan Raaum, Christian Roos
ABSTRACT
It is difficult to predict how current climate change will affect wildlife species adapted to a tropical rainforest environment. Understanding how population dynamics fluctuated in such species throughout periods of past climatic change can provide insight into this issue. The drill (Mandrillus leucophaeus) is a large-bodied rainforest adapted mammal found in West Central Africa. In the middle of this endangered monkey's geographic range is Lake Barombi Mbo, which has a well-documented palynological record of environmental change that dates to the Late Pleistocene. We used a Bayesian coalescent-based framework to analyze 2,076 base pairs of mitochondrial DNA across wild drill populations to infer past changes in female effective population size since the Late Pleistocene. Our results suggest that the drill underwent a nearly 15-fold demographic collapse in female effective population size that was most prominent during the Mid Holocene (approximately 3-5 Ka). This time period coincides with a period of increased dryness and seasonality across Africa and a dramatic reduction in forest coverage at Lake Barombi Mbo. We believe that these changes in climate and forest coverage were the driving forces behind the drill population decline. Furthermore, the warm temperatures and increased aridity of the Mid Holocene are potentially analogous to current and future conditions faced by many tropical rainforest communities. In order to prevent future declines in population size in rainforest-adapted species such as the drill, large tracts of forest should be protected to both preserve habitat and prevent forest loss through aridification.
Product Description
Genomiphi Hy Kit, Cytiva 25-6600-25, sufficient for 1000 reactions
GenomiPhi HY Kit, Cytiva 25-6600-22, sufficient for 25 reactions
Genomiphi Hy Kit, Cytiva 25-6600-20, sufficient for 100 reactions