Merck
  • Injection of autologous bone marrow cells in hyaluronan hydrogel improves cardiac performance after infarction in pigs.
Injection of autologous bone marrow cells in hyaluronan hydrogel improves cardiac performance after infarction in pigs.
American journal of physiology. Heart and circulatory physiology (2014-02-11)
Chien-Hsi Chen, Ming-Yao Chang, Shoei-Shen Wang, Patrick C H Hsieh
ABSTRACT
Intramyocardial injection of bone marrow mononuclear cells (MNCs) with hyaluronan (HA) hydrogel is beneficial to the ischemic heart in a rat model of myocardial infarction (MI). However, the therapeutic efficacy and safety must be addressed in large animals before moving onto a clinical trial. Therefore, the effect of combined treatment on MI was investigated in pigs. Coronary artery ligation was performed in minipigs to induce MI followed by an intramyocardial injection of normal saline (n = 7), HA (n = 7), normal saline with 1 × 10(8) freshly isolated MNCs (n = 8), or HA with 1 × 10(8) MNCs (HA-MNC; n = 7), with a sham-operated group serving as a control (n = 7). The response of each experimental group was estimated by echocardiography, ventricular catheterization, and histological analysis. Although injection of HA or MNCs slightly elevated left ventricular ejection fraction, the combined HA-MNC injection showed a significant increase in left ventricular ejection fraction, contractility, infarct size, and neovascularization. Importantly, injection of MNCs with HA also promoted MNC retention and MNC differentiation into vascular lineage cells in pigs. Therefore, this study not only provides evidence but also raises the possibility of using a combined HA-MNC injection as a promising therapy for heart repair.
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,750,000-2,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor