Merck
  • Menopausal hormone therapy and health outcomes during the intervention and extended poststopping phases of the Women's Health Initiative randomized trials.
Menopausal hormone therapy and health outcomes during the intervention and extended poststopping phases of the Women's Health Initiative randomized trials.
JAMA (2013-10-03)
JoAnn E Manson, Rowan T Chlebowski, Marcia L Stefanick, Aaron K Aragaki, Jacques E Rossouw, Ross L Prentice, Garnet Anderson, Barbara V Howard, Cynthia A Thomson, Andrea Z LaCroix, Jean Wactawski-Wende, Rebecca D Jackson, Marian Limacher, Karen L Margolis, Sylvia Wassertheil-Smoller, Shirley A Beresford, Jane A Cauley, Charles B Eaton, Margery Gass, Judith Hsia, Karen C Johnson, Charles Kooperberg, Lewis H Kuller, Cora E Lewis, Simin Liu, Lisa W Martin, Judith K Ockene, Mary Jo O'Sullivan, Lynda H Powell, Michael S Simon, Linda Van Horn, Mara Z Vitolins, Robert B Wallace
ABSTRACT
Menopausal hormone therapy continues in clinical use but questions remain regarding its risks and benefits for chronic disease prevention. To report a comprehensive, integrated overview of findings from the 2 Women's Health Initiative (WHI) hormone therapy trials with extended postintervention follow-up. A total of 27,347 postmenopausal women aged 50 to 79 years were enrolled at 40 US centers. Women with an intact uterus received conjugated equine estrogens (CEE; 0.625 mg/d) plus medroxyprogesterone acetate (MPA; 2.5 mg/d) (n = 8506) or placebo (n = 8102). Women with prior hysterectomy received CEE alone (0.625 mg/d) (n = 5310) or placebo (n = 5429). The intervention lasted a median of 5.6 years in CEE plus MPA trial and 7.2 years in CEE alone trial with 13 years of cumulative follow-up until September 30, 2010. Primary efficacy and safety outcomes were coronary heart disease (CHD) and invasive breast cancer, respectively. A global index also included stroke, pulmonary embolism, colorectal cancer, endometrial cancer, hip fracture, and death. During the CEE plus MPA intervention phase, the numbers of CHD cases were 196 for CEE plus MPA vs 159 for placebo (hazard ratio [HR], 1.18; 95% CI, 0.95-1.45) and 206 vs 155, respectively, for invasive breast cancer (HR, 1.24; 95% CI, 1.01-1.53). Other risks included increased stroke, pulmonary embolism, dementia (in women aged ≥65 years), gallbladder disease, and urinary incontinence; benefits included decreased hip fractures, diabetes, and vasomotor symptoms. Most risks and benefits dissipated postintervention, although some elevation in breast cancer risk persisted during cumulative follow-up (434 cases for CEE plus MPA vs 323 for placebo; HR, 1.28 [95% CI, 1.11-1.48]). The risks and benefits were more balanced during the CEE alone intervention with 204 CHD cases for CEE alone vs 222 cases for placebo (HR, 0.94; 95% CI, 0.78-1.14) and 104 vs 135, respectively, for invasive breast cancer (HR, 0.79; 95% CI, 0.61-1.02); cumulatively, there were 168 vs 216, respectively, cases of breast cancer diagnosed (HR, 0.79; 95% CI, 0.65-0.97). Results for other outcomes were similar to CEE plus MPA. Neither regimen affected all-cause mortality. For CEE alone, younger women (aged 50-59 years) had more favorable results for all-cause mortality, myocardial infarction, and the global index (nominal P < .05 for trend by age). Absolute risks of adverse events (measured by the global index) per 10,000 women annually taking CEE plus MPA ranged from 12 excess cases for ages of 50-59 years to 38 for ages of 70-79 years; for women taking CEE alone, from 19 fewer cases for ages of 50-59 years to 51 excess cases for ages of 70-79 years. Quality-of-life outcomes had mixed results in both trials. Menopausal hormone therapy has a complex pattern of risks and benefits. Findings from the intervention and extended postintervention follow-up of the 2 WHI hormone therapy trials do not support use of this therapy for chronic disease prevention, although it is appropriate for symptom management in some women. clinicaltrials.gov Identifier: NCT00000611.
Product Description
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Pricing and availability is not currently available.
Medroxyprogesterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.