Merck
Cerebral organoids model human brain development and microcephaly.
Nature (2013-09-03)
Madeline A Lancaster, Magdalena Renner, Carol-Anne Martin, Daniel Wenzel, Louise S Bicknell, Matthew E Hurles, Tessa Homfray, Josef M Penninger, Andrew P Jackson, Juergen A Knoblich
ABSTRACT
The complexity of the human brain has made it difficult to study many brain disorders in model organisms, highlighting the need for an in vitro model of human brain development. Here we have developed a human pluripotent stem cell-derived three-dimensional organoid culture system, termed cerebral organoids, that develop various discrete, although interdependent, brain regions. These include a cerebral cortex containing progenitor populations that organize and produce mature cortical neuron subtypes. Furthermore, cerebral organoids are shown to recapitulate features of human cortical development, namely characteristic progenitor zone organization with abundant outer radial glial stem cells. Finally, we use RNA interference and patient-specific induced pluripotent stem cells to model microcephaly, a disorder that has been difficult to recapitulate in mice. We demonstrate premature neuronal differentiation in patient organoids, a defect that could help to explain the disease phenotype. Together, these data show that three-dimensional organoids can recapitulate development and disease even in this most complex human tissue.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-TSH2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Prox1 Antibody, clone 4G10, clone 4G10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-189 mreelin, clone 142, clone 142, Chemicon®, from mouse