Merck
  • Comparison of three buffers used in the formulation of buffered charcoal yeast extract medium.
Comparison of three buffers used in the formulation of buffered charcoal yeast extract medium.
Journal of clinical microbiology (1993-12-01)
P H Edelstein, M A Edelstein
ABSTRACT
Growth of Legionella spp. on buffered charcoal yeast extract medium supplemented with alpha-ketoglutarate and formulated with 3-(n-morpholino)propanesulfonic acid (MOPS), 3-(n-morpholino)-2-hydroxypropanesulfonic acid (MOPSO), or n-(2-acetamido)-2-aminoethanesulfonic acid (ACES) buffer was similar. With three exceptions, growth was no different in buffered yeast extract broth supplemented with alpha-ketoglutarate and formulated with MOPS or ACES buffer.
Sigma-Aldrich
ACES, BioPerformance Certified, ≥99.0%, suitable for cell culture
Sigma-Aldrich
ACES, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
ACES, ≥99.0% (titration)
Sigma-Aldrich
ACES, BioUltra, ≥99.5% (T)