Merck
  • [The stereoselectivity of chrono-pharmacokinetic parameters of hydratropic acid in rats].
[The stereoselectivity of chrono-pharmacokinetic parameters of hydratropic acid in rats].
Yao xue xue bao = Acta pharmaceutica Sinica (1993-01-01)
Y S He, H L Lu, D Y Wang, S X He
ABSTRACT
The stereoselective chrono-pharmacokinetic parameters of hydratropic acid in rats were studied. The results showed that under standard light-dark cycle pharmacokinetic parameters of T1/2 alpha and CL are stereoselective and under reverse light-dark cycle, parameters T1/2 beta, AUC, CL, Vc and MRT were shown to be stereoselective. To compare the corresponding parameters of the two different light-dark cycles using t-test, no differences were found in most of them.
Sigma-Aldrich
(S)-(+)-2-Phenylpropionic acid, 97%
Sigma-Aldrich
2-Phenylpropionic acid, 97%