Merck
  • The effects of homologous series of anaesthetics on a resting potassium conductance of the squid giant axon.
The effects of homologous series of anaesthetics on a resting potassium conductance of the squid giant axon.
Biochimica et biophysica acta (1989-01-30)
A A Elliott, J R Elliott, D A Haydon
ABSTRACT
The effects of n-alkanes (n-pentane to n-octane), n-alkanols (n-pentanol to n-undecanol) and two carboxylic esters (methyl pentanoate and methyl octanoate) on the conductance of squid giant axons in a high potassium, zero sodium bathing solution have been examined. Sodium and delayed rectifier potassium channels were as far as possible pharmacologically blocked. A substantial fraction of the measured conductance is attributed to a recently-described, voltage-independent, potassium channel. Anaesthetics block this channel but its sensitivity is markedly different from those of other squid axon ion channels.
Sigma-Aldrich
Methyl octanoate, 99%, FG
Supelco
Methyl octanoate, analytical standard
Sigma-Aldrich
Methyl octanoate, 99%