Merck
  • Immobilization of iron oxide nanoclusters on surface functionalized silicon substrate and their catalytic behavior to synthesize multi-walled carbon nanotubes.
Immobilization of iron oxide nanoclusters on surface functionalized silicon substrate and their catalytic behavior to synthesize multi-walled carbon nanotubes.
Journal of nanoscience and nanotechnology (2013-08-02)
Prashanta Dhoj Adhikari, Sunghwan Kim, Sui Lee, Chong-Yun Park
ABSTRACT
Multi-walled carbon nanotubes (MWCNTs) were grown using iron oxide nanoclusters (Nc) assisted by self-assembled monolayer (SAM) on substrate. The amine-terminated SAM fabricated on silicon substrate (APTMS/SI) was carried out by UV-treatment and immersed into the FeCI3/HCI aqueous solution. Then, Nc were immobilized onto oxidized SAM silicon substrate (SAMs/Si) through electrostatic interaction between cationic Nc and anionic SAMs/Si. The characterization results clearly show that the well-graphitized MWCNTs were synthesized by using functionalized silicon substrate (Nc/SAMs/Si) as a template having appropriate density of catalyst. These consequences show that SAM containing template is important to achieve the effective layer of catalyst to synthesize MWCNTs in chemical vapor deposition (CVD).
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Iron(III) oxide, nanopowder, <50 nm particle size (BET)
Sigma-Aldrich
Iron(III) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Iron(III) oxide, powder, <5 μm, ≥96%
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol