Merck
  • Effects of surface morphology of ZnO seed layers on growth of ZnO nanostructures prepared by hydrothermal method and annealing.
Effects of surface morphology of ZnO seed layers on growth of ZnO nanostructures prepared by hydrothermal method and annealing.
Journal of nanoscience and nanotechnology (2013-07-19)
Kwang Gug Yim, Min Su Kim, Jae-Young Leem
ABSTRACT
ZnO nanostructures were grown on Si (111) substrates by a hydrothermal method. Prior to growing the ZnO nanostructures, ZnO seed layers with different post-heat temperatures were prepared by a spin-coating process. Then, the ZnO nanostructures were annealed at 500 degrees C for 20 min under an Ar atmosphere. Scanning electron microscopy (SEM), X-ray diffraction (XRD), and photoluminescence (PL) were carried out at room temperature (RT) to investigate the structural and optical properties of the as-grown and annealed ZnO nanostructures. The surface morphologies of the seed layers changed from a smooth surface to a mountain chain-like structure as the post-heating temperatures increased. The as-grown and annealed ZnO nanostructures exhibited a strong (002) diffraction peak. Compared to the as-grown ZnO nanostructures, the annealed ZnO nanostructures exhibited significantly strong enhancement in the PL intensity ratio by almost a factor of 2.
Product Description
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)