Merck
  • Salivary trace elements in relation to dental caries in a group of multi-ethnic schoolchildren in Shah Alam, Malaysia.
Salivary trace elements in relation to dental caries in a group of multi-ethnic schoolchildren in Shah Alam, Malaysia.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry (2013-06-14)
A S Hussein, H F Ghasheer, N M Ramli, R J Schroth, M I Abu-Hassan
ABSTRACT
To assess the salivary levels of Copper (Cu), Zinc (Zn), Manganese (Mn) and Iron (Fe) obtained from children of different ethnic backgrounds in Shah Alam, Malaysia and investigate the possible relationships with caries. One hundred and twenty primary school children were included. They were divided into caries and caries-free groups. Unstimulated whole saliva was collected from each participant using spitting method. The salivary elements were measured using an Atomic Absorption Spectrophotometer. Descriptive statistics, bivariate and Pearson's correlation analysis were performed. Salivary Cu and Zn levels were significantly higher in children with dental caries compared to those caries-free (p < 0.05). Moreover, these elements had a positive correlation with dental caries (Cu: r=0.698, p<0.001; Zn: r=0.181, p<0.05). No significant variations in Mn and Fe were observed between caries and caries-free group (p>0.05). Additionally, there were significant differences in salivary Zn and Fe among different age groups (p<0.05) and highly significant differences in salivary Cu, Mn and Fe among different ethnic groups (p<0.001). However, all elements exhibited no significant differences between males and females. The salivary Cu and Zn levels showed significant differences between caries and caries-free groups. The findings also revealed significant variations in the levels of salivary Cu, Mn and Fe among different ethnic groups and salivary Zn and Fe among different age groups.
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis