Merck
  • Brain mitochondrial metabolic dysfunction and glutamate level reduction in the pilocarpine model of temporal lobe epilepsy in mice.
Brain mitochondrial metabolic dysfunction and glutamate level reduction in the pilocarpine model of temporal lobe epilepsy in mice.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2013-04-25)
Olav B Smeland, Mussie G Hadera, Tanya S McDonald, Ursula Sonnewald, Karin Borges
ABSTRACT
Although certain metabolic characteristics such as interictal glucose hypometabolism are well established for temporal lobe epilepsy (TLE), its pathogenesis still remains unclear. Here, we performed a comprehensive study of brain metabolism in a mouse model of TLE, induced by pilocarpine-status epilepticus (SE). To investigate glucose metabolism, we injected mice 3.5-4 weeks after SE with [1,2-(13)C]glucose before microwave fixation of the head. Using (1)H and (13)C nuclear magnetic resonance spectroscopy, gas chromatography-mass spectrometry and high-pressure liquid chromatography, we quantified metabolites and (13)C labeling in extracts of cortex and hippocampal formation (HF). Hippocampal levels of glutamate, glutathione and alanine were decreased in pilocarpine-SE mice compared with controls. Moreover, the contents of N-acetyl aspartate, succinate and reduced nicotinamide adenine dinucleotide (phosphate) NAD(P)H were decreased in HF indicating impairment of mitochondrial function. In addition, the reduction in (13)C enrichment of hippocampal citrate and malate suggests decreased tricarboxylic acid (TCA) cycle turnover in this region. In cortex, we found reduced (13)C labeling of glutamate, glutamine and aspartate via the pyruvate carboxylation and pyruvate dehydrogenation pathways, suggesting slower turnover of these amino acids and/or the TCA cycle. In conclusion, mitochondrial metabolic dysfunction and altered amino-acid metabolism is found in both cortex and HF in this epilepsy model.
Sigma-Aldrich
Pilocarpine nitrate, meets USP testing specifications
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine nitrate salt, ≥98% (HPLC)
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Pilocarpine nitrate for system suitability, European Pharmacopoeia (EP) Reference Standard