Merck
  • Mesoporous silica nanoparticle facilitated drug release through cascade photosensitizer activation and cleavage of singlet oxygen sensitive linker.
Mesoporous silica nanoparticle facilitated drug release through cascade photosensitizer activation and cleavage of singlet oxygen sensitive linker.
Chemical communications (Cambridge, England) (2013-01-18)
Junseok Lee, Juhee Park, Kaushik Singha, Won Jong Kim
ABSTRACT
A photoresponsive drug releasing system based on mesoporous silica nanoparticles (MSN) was developed. A model drug was decorated on the surfaces via singlet oxygen sensitive linkers and a singlet oxygen-generating photosensitizer was loaded in the mesopores of the MSN. An on-demand drug releasing system was successfully developed by irradiation with long-wavelength light.
Sigma-Aldrich
Zinc phthalocyanine, Dye content 97 %