Merck
  • Sprague-Dawley rats bearing McA-RH7777 cells for study of hepatoma and transarterial chemoembolization.
Sprague-Dawley rats bearing McA-RH7777 cells for study of hepatoma and transarterial chemoembolization.
Anticancer research (2012-12-26)
Hye Rim Cho, Jin Woo Choi, Hyo-Cheol Kim, Yong Sub Song, Gyoung Min Kim, Kyu Ri Son, Jin Wook Chung
ABSTRACT
To evaluate the feasibility of the McA-RH7777 tumor model in Sprague-Dawley (SD) rats, for study of hepatoma and transarterial chemoembolization. McA-RH7777 rat hepatoma cells (1×10(7)) were inoculated into the left hepatic lobe of SD rats (n=38). Chemoembolization with left common carotid artery access was performed using an emulsion of iodized oil and doxorubicin, and polyvinyl alcohol particles. Tumor induction rate and response to chemoembolization were assessed by magnetic resonance imaging and histology. Tumor induction rate of McA-RH7777 in SD rat livers was 73.3% (11/15). Hematoxylin-and-eosin staining revealed hypercellular tumor with a trabecular pattern that mimics human hepatocellular carcinoma. Chemoembolization was successfully conducted in all rats. There was a significant difference in tumor growth rates between the chemoembolization-treated and control groups (p<0.0001). A rat tumor model of McA-RH7777 cells in SD rats is feasible and has the potential to be a good model for hepatoma and chemoembolization studies.
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000