Merck
  • Amphiphile behavior in mixed solvent media I: self-aggregation and ion association of sodium dodecylsulfate in 1,4-dioxane-water and methanol-water media.
Amphiphile behavior in mixed solvent media I: self-aggregation and ion association of sodium dodecylsulfate in 1,4-dioxane-water and methanol-water media.
Langmuir : the ACS journal of surfaces and colloids (2012-09-07)
A Pan, B Naskar, G K S Prameela, B V N Phani Kumar, A B Mandal, S C Bhattacharya, S P Moulik
ABSTRACT
Mixed aquo-organic solvents are used in chemical, industrial, and pharmaceutical processes along with amphiphilic materials. Their fundamental studies with reference to bulk and interfacial phenomena are thus considered to be important, but such detailed studies are limited. In this work, the interfacial adsorption of sodium dodecylsulfate (SDS, C12H25SO4(-)Na(+)) in dioxane-water (Dn-W) and methanol-water (Ml-W) media in extensive mixing ratios along with its bulk behavior have been investigated. The solvent-composition-dependent properties have been identified, and their quantifications have been attempted. The SDS micellization has been assessed in terms of different solvent parameters, and the possible formation of an ion pair and triple ion of the colloidal electrolyte, C12H25SO4(-)Na(+) in the Dn-W medium has been correlated and quantified. In the Ml-W medium at a high volume percent of Ml, the SDS amphiphile formed special associated species instead of ion association. The formation of self-assembly and the energetics of SDS in the mixed solvent media have been determined and assessed using conductometry, calorimetry, tensiometry, viscometry, NMR, and DLS methods. The detailed study undertaken herein with respect to the behavior of SDS in the mixed aquo-organic solvent media (Dn-W and Ml-W) is a new kind of endeavor.
Product Description
Supelco
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
Supelco
1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D)
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
Supelco
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
Supelco
1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, ReagentPlus®, ≥99%, contains ≤25 ppm BHT as stabilizer