Merck
  • Improvement of dissolution and hypoglycemic efficacy of glimepiride by different carriers.
Improvement of dissolution and hypoglycemic efficacy of glimepiride by different carriers.
AAPS PharmSciTech (2012-07-31)
Elham A Mohamed, Mahasen M Meshali, Abdel Monem M Foda, Thanaa M Borg
ABSTRACT
Effects of tromethamine (Tris), polyvinylpyrrolidone (PVP-K25), and low molecular weight chitosan (LM-CH) on dissolution and therapeutic efficacy of glimepiride (Gmp) were investigated using physical mixtures (PMs), coground mixtures, coprecipitates (Coppts) or kneaded mixtures (KMs), and compared with drug alone. Fourier transform infrared spectroscopy, differential scanning colorimetry, and X-ray diffractometry were performed to identify any physicochemical interaction with Gmp. Surface morphology was examined via scanning electron microscopy. The results of Gmp in vitro dissolution revealed that it was greatly enhanced by Coppt with Tris or PVP-K25 and KM with LM-CH at a drug to carrier ratio of 1:8. Gmp amorphization by PVP-K25 and LM-CH was a major factor in increasing Gmp dissolution. Being basic, Tris might increase the pH of the microdiffusion layer around Gmp particles improving its dissolution. Formation of water-soluble complexes suggested by solubility study may also explain the enhanced dissolution. Capsules were prepared from Coppts and KM 1:8 drug to carrier binary systems and also with Tris PMs. In vivo, the hypoglycemic efficacy of Gmp capsules in rabbits increased by 1.63-, 1.50-, and 1.46-fold for 1:8 Coppts with Tris or PVP-K25 and KM with LM-CH respectively, compared with Gmp alone. Surprisingly, the response to Tris PM 1:20 capsules was 1.52-fold revealing statistically insignificant difference to that of Tris Coppt 1:8 (1.63 fold). As a conclusion, dissolution enhancement and hypoglycemic potentiation by 1:20 PM of Gmp/Tris, being simple and easy to prepare, may enable development of a reduced-dose and fast-release oral dosage form of Gmp.
Product Description
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid