Merck
  • Inactivation of a peroxiredoxin by hydrogen peroxide is critical for thioredoxin-mediated repair of oxidized proteins and cell survival.
Inactivation of a peroxiredoxin by hydrogen peroxide is critical for thioredoxin-mediated repair of oxidized proteins and cell survival.
Molecular cell (2012-01-17)
Alison M Day, Jonathon D Brown, Sarah R Taylor, Jonathan D Rand, Brian A Morgan, Elizabeth A Veal
ABSTRACT
Eukaryotic 2-Cys peroxiredoxins (Prx) are abundant antioxidant enzymes whose thioredoxin peroxidase activity plays an important role in protecting against oxidative stress, aging, and cancer. Paradoxically, this thioredoxin peroxidase activity is highly sensitive to inactivation by peroxide-induced Prx hyperoxidation. However, any possible advantage in preventing Prx from removing peroxides under oxidative stress conditions has remained obscure. Here we demonstrate that, in cells treated with hydrogen peroxide, the Prx Tpx1 is a major substrate for thioredoxin in the fission yeast Schizosaccharomyces pombe and, as such, competitively inhibits thioredoxin-mediated reduction of other oxidized proteins. Consequently, we reveal that the hyperoxidation of Tpx1 is critical to allow thioredoxin to act on other substrates ensuring repair of oxidized proteins and cell survival following exposure to toxic levels of hydrogen peroxide. We conclude that the inactivation of the thioredoxin peroxidase activity of Prx is important to maintain thioredoxin activity and cell viability under oxidative stress conditions.
Sigma-Aldrich
DL-Methionine sulfoxide, ≥98.5% (NT)
Sigma-Aldrich
L-Methionine sulfoxide