Merck
  • Estimation and characterization of PCDD/Fs, dl-PCBs, PCNs, HxCBz and PeCBz emissions from magnesium metallurgy facilities in China.
Estimation and characterization of PCDD/Fs, dl-PCBs, PCNs, HxCBz and PeCBz emissions from magnesium metallurgy facilities in China.
Chemosphere (2011-10-26)
Zhiqiang Nie, Minghui Zheng, Wenbin Liu, Bing Zhang, Guorui Liu, Guijin Su, Pu Lv, Ke Xiao
ABSTRACT
Magnesium production is considered to be one potential source of unintentional persistent organic pollutants (unintentional POPs). However, studies on the emissions of unintentional POPs from magnesium metallurgy are still lacking. Emissions of unintentional POPs, such as polychlorinated dibenzo-p-dioxins/dibenzofurans (PCDD/Fs), dioxin-like polychlorinated biphenyls (dl-PCBs), polychlorinated naphthalenes (PCNs), hexachlorobenzene (HxCBz) and pentachlorobenzene (PeCBz) are covered under the Stockholm Convention. In this study, these emissions were investigated through a magnesium smelting process. Stack gas and fly ash samples from a typical magnesium plant in China were collected and analyzed to estimate the emissions of unintentional POPs from magnesium metallurgy. Emissions factors of 412 ng TEQ t(-1) for PCDD/Fs, 18.6 ng TEQ t(-1) for dl-PCBs, 3329 μg t(-1) for PCNs, 820 μg t(-1) for HxCBz, and 1326 μg t(-1) for PeCBz were obtained in 2009. Annual emissions from magnesium metallurgy in China were estimated to be 0.46 g WHO-TEQ for PCDD/Fs and dl-PCBs, 1651 g for PCNs, 403 g for HxCBz and 653 g for PeCBz, respectively.
Product Description
Supelco
Pentachlorobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachlorobenzene, 96%