Merck
  • Characterization of emission properties of Er3+ ions in TeO2-CdF2-WO3 glasses.
Characterization of emission properties of Er3+ ions in TeO2-CdF2-WO3 glasses.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-09-20)
G Bilir, N Mustafaoglu, G Ozen, B DiBartolo
ABSTRACT
TeO(2)-CdF(2)-WO(3) glasses with various compositions and Er(3+) concentrations were prepared by conventional melting method. Their optical properties were studied by measuring the absorption, luminescence spectra and the decay patterns at room temperature. From the optical absorption spectra the Judd-Ofelt parameters (Ω(t)), transition probabilities, branching ratios of various transitions, and radiative lifetimes were calculated. The absorption and emission cross-section spectra of the (4)I(15/2) to (4)I(13/2) transition of erbium were determined. Emission quantum efficiencies and the average critical distance R(0) which provides a measure for the strength of cross relaxation were determined.
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Tellurium dioxide, ≥97.0%
Sigma-Aldrich
Tellurium dioxide, ≥99%