Merck
  • [Forty-four years of advances in pathogenesis, diagnosis and treatment of flatulence syndrome].
[Forty-four years of advances in pathogenesis, diagnosis and treatment of flatulence syndrome].
Revista de gastroenterologia del Peru : organo oficial de la Sociedad de Gastroenterologia del Peru (2010-07-21)
Raúl León Barúa
ABSTRACT
In the present paper, the author reminds investigations performed, since forty-four years ago, to clarify the etiopathogenesis and to improve the diagnosis and treatment of the flatulence syndrome.
Sigma-Aldrich
Bismuth(III) subsalicylate, 99.9% trace metals basis
Pricing and availability is not currently available.