Merck
  • Amplification of anion sensing by disulfide functionalized ferrocene and ferrocene-calixarene receptors adsorbed onto gold surfaces.
Amplification of anion sensing by disulfide functionalized ferrocene and ferrocene-calixarene receptors adsorbed onto gold surfaces.
Dalton transactions (Cambridge, England : 2003) (2010-06-17)
David P Cormode, Andrew J Evans, Jason J Davis, Paul D Beer
ABSTRACT
A disulfide functionalized bis-ferrocene urea acyclic receptor and disulfide functionalized mono- and bis-ferrocene amide and urea appended upper rim calix[4]arene receptors were prepared for the fabrication of SAM redox-active anion sensors. 1H NMR and diffusive voltammetric anion recognition investigations revealed each receptor to be capable of complexing and electrochemically sensing anions via cathodic perturbations of the respective receptor's ferrocene/ferrocenium redox couple. SAMs of a ferrocene urea receptor 3 and ferrocene urea calixarene receptor 17 exhibited significant enhanced magnitudes of cathodic response upon anion addition as compared to observed diffusive perturbations. SAMs of 17 were demonstrated to sense the perrhenate anion in aqueous solutions.
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium perrhenate, ≥99%
Sigma-Aldrich
Ammonium perrhenate, 99.999% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis