Merck
  • Oral absorption and oxidative metabolism of atrazine in rats evaluated by physiological modeling approaches.
Oral absorption and oxidative metabolism of atrazine in rats evaluated by physiological modeling approaches.
Toxicology (2007-09-05)
Tami S McMullin, William H Hanneman, Brian K Cranmer, John D Tessari, Melvin E Andersen
ABSTRACT
Atrazine (ATRA) is metabolized by cytochrome P450s to the chlorinated metabolites, 2-chloro-4-ethylamino-6-amino-1,3,5-triazine (ETHYL), 2-chloro-4-amino-6-isopropylamino-1, 3, 5-triazine (ISO), and diaminochlorotriazine (DACT). Here, we develop a set of physiologically based pharmacokinetic (PBPK) models that describe the influence of oral absorption and oxidative metabolism on the blood time course curves of individual chlorotriazines (Cl-TRIs) in rat after oral dosing of ATRA. These models first incorporated in vitro metabolic parameters to describe time course plasma concentrations of DACT, ETHYL, and ISO after dosing with each compound. Parameters from each individual model were linked together into a final composite model in order to describe the time course of all 4 Cl-TRIs after ATRA dosing. Oral administration of ISO, ETHYL and ATRA produced double peaks of the compounds in plasma time courses that were described by multiple absorption phases from gut. An adequate description of the uptake and bioavailability of absorbed ATRA also required inclusion of additional oxidative metabolic clearance of ATRA to the mono-dealkylated metabolites occurring in GI a tract compartment. These complex processes regulating tissue dosimetry of atrazine and its chlorinated metabolites likely reflect limited compound solubility in the gut from dosing with an emulsion, and sequential absorption and metabolism along the GI tract at these high oral doses.
Product Description
Supelco
Atrazine-desethyl, PESTANAL®, analytical standard
Supelco
Atrazine-desisopropyl, PESTANAL®, analytical standard
Sigma-Aldrich
2-Chloro-4,6-diamino-1,3,5-triazine, 95%