Merck
  • Hyphenation of Raman spectroscopy with gravimetric analysis to interrogate water-solid interactions in pharmaceutical systems.
Hyphenation of Raman spectroscopy with gravimetric analysis to interrogate water-solid interactions in pharmaceutical systems.
Journal of pharmaceutical and biomedical analysis (2006-08-03)
Alan D Gift, Lynne S Taylor
ABSTRACT
A moisture sorption gravimetric analyzer has been combined with a Raman spectrometer to better understand the various modes of water-solid interactions relevant to pharmaceutical systems. A commercial automated moisture sorption balance was modified to allow non-contact monitoring of the sample properties by interfacing a Raman probe with the sample holder. This hybrid instrument allows for gravimetric and spectroscopic changes to be monitored simultaneously. The utility of this instrument was demonstrated by investigating different types of water-solid interactions including stoichiometric and non-stoichiometric hydrate formation, deliquescence, amorphous-crystalline transformation, and capillary condensation. In each of the model systems, sulfaguanidine, cromolyn sodium, ranitidine HCl, amorphous sucrose and silica gel, spectroscopic changes were observed during the time course of the moisture sorption profile. Analysis of spectroscopic data provided information about the origin of the observed changes in moisture content as a function of relative humidity. Furthermore, multivariate data analysis techniques were employed as a means of processing the spectroscopic data. Principle components analysis was found to be useful to aid in data processing, handling and interpretation of the spectral changes that occurred during the time course of the moisture sorption profile.
Sigma-Aldrich
Sulfaguanidine
Supelco
Sulfaguanidine, VETRANAL®, analytical standard