Merck
  • NAD(P)-dependent aldehyde dehydrogenases induced during growth of Ralstonia eutropha strain Bo on tetrahydrofurfuryl alcohol.
NAD(P)-dependent aldehyde dehydrogenases induced during growth of Ralstonia eutropha strain Bo on tetrahydrofurfuryl alcohol.
Journal of bacteriology (2001-11-22)
T Schräder, G Zarnt, J R Andreesen
ABSTRACT
Different aldehyde dehydrogenases (AlDHs) were formed during growth of Ralstonia eutropha Bo on tetrahydrofurfuryl alcohol (THFA). One of these enzymes, AlDH 4, was purified and characterized as a homodimer containing no prosthetic groups, showing a strong substrate inhibition, and having an N-terminal sequence similar to those of various NAD(P)-dependent AlDHs. The conversion rate of THFA by the quinohemoprotein THFA dehydrogenase was increased by AlDH 4.
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 99%
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, 98%
Sigma-Aldrich
Tetrahydrofurfuryl alcohol, ≥98%