Merck
Crystal structure of human prostatic acid phosphatase .
The Prostate (2000-01-19)
C G Jakob, K Lewinski, R Kuciel, W Ostrowski, L Lebioda
ABSTRACT
Prostatic acid phosphatase (hPAP) is a major product of the human prostate gland, yet its physiological substrate remains unknown. Human PAP, purified from semen, was crystallized using polyethylene glycol as the precipitant and its crystal structure was determined using X-ray diffraction. The structure was refined at 3.1 A resolution to R = 16% and R(free) = 27%. The structure of hPAP is similar to that of other known histidine phosphatases, and the positions of its catalytic residues are conserved. N-linked carbohydrates are present at each of the possible glycosylation sites. It appears that high-mannose chains are attached to Asn 62 and Asp 301, while complex chains are at Asn 188. The similarity of the three-dimensional structures of rat PAP and human PAP indicates that the mechanistic analyses of the catalytic mechanism proposed for the rat enzyme should be extended to the human enzyme without reservations. The crystallographic data allowed the correlation of attachment sites of N-linked carbohydrate chains with a given carbohydrate type. The carbohydrates of the protein produced in the prostate cells and in the baculovirus expression system appear to differ at the site of complex carbohydrates attachment.
Sigma-Aldrich
5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid