Merck
  • Histopathologic and proteogenomic heterogeneity reveals features of clear cell renal cell carcinoma aggressiveness.
Histopathologic and proteogenomic heterogeneity reveals features of clear cell renal cell carcinoma aggressiveness.
Cancer cell (2022-12-24)
Yize Li, Tung-Shing M Lih, Saravana M Dhanasekaran, Rahul Mannan, Lijun Chen, Marcin Cieslik, Yige Wu, Rita Jiu-Hsien Lu, David J Clark, Iga Kołodziejczak, Runyu Hong, Siqi Chen, Yanyan Zhao, Seema Chugh, Wagma Caravan, Nataly Naser Al Deen, Noshad Hosseini, Chelsea J Newton, Karsten Krug, Yuanwei Xu, Kyung-Cho Cho, Yingwei Hu, Yuping Zhang, Chandan Kumar-Sinha, Weiping Ma, Anna Calinawan, Matthew A Wyczalkowski, Michael C Wendl, Yuefan Wang, Shenghao Guo, Cissy Zhang, Anne Le, Aniket Dagar, Alex Hopkins, Hanbyul Cho, Felipe da Veiga Leprevost, Xiaojun Jing, Guo Ci Teo, Wenke Liu, Melissa A Reimers, Russell Pachynski, Alexander J Lazar, Arul M Chinnaiyan, Brian A Van Tine, Bing Zhang, Karin D Rodland, Gad Getz, D R Mani, Pei Wang, Feng Chen, Galen Hostetter, Mathangi Thiagarajan, W Marston Linehan, David Fenyö, Scott D Jewell, Gilbert S Omenn, Rohit Mehra, Maciej Wiznerowicz, Ana I Robles, Mehdi Mesri, Tara Hiltke, Eunkyung An, Henry Rodriguez, Daniel W Chan, Christopher J Ricketts, Alexey I Nesvizhskii, Hui Zhang, Li Ding
ABSTRACT
Clear cell renal cell carcinomas (ccRCCs) represent ∼75% of RCC cases and account for most RCC-associated deaths. Inter- and intratumoral heterogeneity (ITH) results in varying prognosis and treatment outcomes. To obtain the most comprehensive profile of ccRCC, we perform integrative histopathologic, proteogenomic, and metabolomic analyses on 305 ccRCC tumor segments and 166 paired adjacent normal tissues from 213 cases. Combining histologic and molecular profiles reveals ITH in 90% of ccRCCs, with 50% demonstrating immune signature heterogeneity. High tumor grade, along with BAP1 mutation, genome instability, increased hypermethylation, and a specific protein glycosylation signature define a high-risk disease subset, where UCHL1 expression displays prognostic value. Single-nuclei RNA sequencing of the adverse sarcomatoid and rhabdoid phenotypes uncover gene signatures and potential insights into tumor evolution. In vitro cell line studies confirm the potential of inhibiting identified phosphoproteome targets. This study molecularly stratifies aggressive histopathologic subtypes that may inform more effective treatment strategies.
Sigma-Aldrich
Anti-UCHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
UCH-L1 Inhibitor, The UCH-L1 Inhibitor, also referenced under CAS 668467-91-2, controls the biological activity of UCH-L1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride solution, ~0.1 M in ethanol (T)