Merck
  • Discovery of (S)-N-[2-[1-(3-ethoxy-4-methoxyphenyl)-2-methanesulfonylethyl]-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl] acetamide (apremilast), a potent and orally active phosphodiesterase 4 and tumor necrosis factor-alpha inhibitor.
Discovery of (S)-N-[2-[1-(3-ethoxy-4-methoxyphenyl)-2-methanesulfonylethyl]-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl] acetamide (apremilast), a potent and orally active phosphodiesterase 4 and tumor necrosis factor-alpha inhibitor.
Journal of medicinal chemistry (2009-03-05)
Hon-Wah Man, Peter Schafer, Lu Min Wong, Rebecca T Patterson, Laura G Corral, Heather Raymon, Kate Blease, Jim Leisten, Michael A Shirley, Yang Tang, Darius M Babusis, Roger Chen, Dave Stirling, George W Muller
ABSTRACT
In this communication, we report the discovery of 1S (apremilast), a novel potent and orally active phosphodiesterase 4 (PDE4) and tumor necrosis factor-alpha inhibitor. The optimization of previously reported 3-(1,3-dioxo-1,3-dihydroisoindol-2-yl)-3-(3,4-dimethoxyphenyl)propionic acid PDE4 inhibitors led to this series of sulfone analogues. Evaluation of the structure-activity relationship of substitutions on the phthalimide group led to the discovery of an acetylamino analogue 1S, which is currently in clinical trials.
Sigma-Aldrich
Apremilast, ≥98% (HPLC)