Merck
  • A major deletion in the surfactant protein-B gene causing lethal respiratory distress.
A major deletion in the surfactant protein-B gene causing lethal respiratory distress.
Acta paediatrica (Oslo, Norway : 1992) (2007-03-30)
Daniel J Wegner, Torbjörn Hertzberg, Hillary B Heins, Göran Elmberger, Michael J MacCoss, Christopher S Carlson, Lawrence M Nogee, F Sessions Cole, Aaron Hamvas
ABSTRACT
Loss of function mutations in the surfactant protein-B gene (SFTPB) cause lethal neonatal respiratory distress due to reduced or absent expression of mature surfactant protein B (SP-B, encoded in exons 6 and 7). No large deletions in SFTPB have been previously identified. Genomic, proteomic and immunohistochemical characterization of a 3 kb deletion in SFTPB. A full-term newborn presented with refractory respiratory failure. We amplified and sequenced SFTPB from the infant and both parents, determined SP-B protein expression in tracheal aspirate samples using Western-blot analysis, and performed immunohistochemical staining and electron microscopy of lung biopsy tissue. The infant was homozygous for a 2958 bp deletion in SFTPB that included exons 7 and 8. Both asymptomatic parents were heterozygous for the deletion. A truncated mature SP-B peptide was detected on Western blotting of tracheal aspirate. Amino acid sequence specific to that encoded in exon 5 was present, but that encoded by exon 7 was absent. ProSP-B expression was robust within alveolar type II cells and lamellar body structure was disrupted. This deletion in SFTPB resulted in SP-B deficiency due to absence of elements in mature SP-B that are critical for appropriate peptide folding, trafficking and processing.
Sigma-Aldrich
Anti-Prosurfactant Protein B Antibody, reacts with both CT and NT, serum, Chemicon®
Sigma-Aldrich
Anti-Prosurfactant Protein C (proSP-C) Antibody, serum, Chemicon®