Merck
  • Sodium hydroxide pretreatment and enzymatic hydrolysis of coastal Bermuda grass.
Sodium hydroxide pretreatment and enzymatic hydrolysis of coastal Bermuda grass.
Bioresource technology (2010-01-22)
Ziyu Wang, Deepak R Keshwani, Arthur P Redding, Jay J Cheng
ABSTRACT
Coastal Bermuda grass was pretreated with NaOH at concentrations from 0.5% to 3% (w/v) for a residence time from 15 to 90min at 121 degrees C. The pretreatments were evaluated based on total lignin removal and production of total reducing sugars, glucose and xylose from enzymatic hydrolysis of the pretreated biomass. Up to 86% lignin removal was observed. The optimal NaOH pretreatment conditions at 121 degrees C for total reducing sugars production as well as glucose and xylose yields are 15min and 0.75% NaOH. Under these optimal pretreatment conditions, total reducing sugars yield was about 71% of the theoretical maximum, and the overall conversion efficiencies for glucan and xylan were 90.43% and 65.11%, respectively.
Sigma-Aldrich
Sodium hydroxide, BioUltra, ≥98.0% (T), pellets