Merck
  • The Sweet Taste of Acarbose and Maltotriose: Relative Detection and Underlying Mechanism.
The Sweet Taste of Acarbose and Maltotriose: Relative Detection and Underlying Mechanism.
Chemical senses (2018-12-28)
Alexa J Pullicin, Michael H Penner, Juyun Lim
ABSTRACT
Although sweet-tasting saccharides possess similar molecular structures, their relative sweetness often varies to a considerable degree. Current understanding of saccharide structure/sweetness interrelationships is limited. Understanding how certain structural features of saccharides and/or saccharide analogs correlate to their relative sweetness can provide insight on the mechanisms underlying sweetness potency. Maltotriose is a short-chain glucose-based oligosaccharide, which we recently reported to elicit sweet taste. Acarbose, an α-glucosidase inhibitor, is a pseudo-saccharide that has an overall resemblance to a glucose-based oligosaccharide and thus may be viewed as a structural analog. During other studies, we recognized that acarbose can also elicit sweet taste. Here, we formally investigated the underlying taste detection mechanism of acarbose, while confirming our previous findings for maltotriose. We found that subjects could detect the sweet taste of acarbose and maltotriose in aqueous solutions but were not able to detect them in the presence of a sweet taste inhibitor lactisole. These findings support that both are ligands of the human sweet taste receptor, hT1R2/hT1R3. In a separate experiment, we measured the relative sweetness detection of acarbose, maltotriose, and other sweet-tasting mono- and disaccharides (glucose, fructose, maltose, and sucrose). Whereas maltotriose was found to have a similar discriminability profile to glucose and maltose, the discriminability of acarbose matched that of fructose at the concentrations tested (18, 32, and 56 mM). These findings are discussed in terms of how specific molecular features (e.g., degree of polymerization and monomer composition) may contribute to the relative sweetness of saccharides.
Sigma-Aldrich
Maltotriose, ≥90% (HPLC)