Merck
  • TMEM106B deficiency impairs cerebellar myelination and synaptic integrity with Purkinje cell loss.
TMEM106B deficiency impairs cerebellar myelination and synaptic integrity with Purkinje cell loss.
Acta neuropathologica communications (2022-03-16)
Tuancheng Feng, Lin Luan, Isabel Iscol Katz, Mohammed Ullah, Vivianna M Van Deerlin, John Q Trojanowski, Edward B Lee, Fenghua Hu
ABSTRACT
TMEM106B, a type II lysosomal transmembrane protein, has recently been associated with brain aging, hypomyelinating leukodystrophy, frontotemporal lobar degeneration (FTLD) and several other brain disorders. TMEM106B is critical for proper lysosomal function and TMEM106B deficiency leads to myelination defects, FTLD related pathology, and motor coordination deficits in mice. However, the physiological and pathological functions of TMEM106B in the brain are still not well understood. In this study, we investigate the role of TMEM106B in the cerebellum, dysfunction of which has been associated with FTLD and other brain disorders. We found that TMEM106B is ubiquitously expressed in neurons in the cerebellum, with the highest levels in the Purkinje neurons. Aged TMEM106B-deficient mice show significant loss of Purkinje neurons specifically in the anterior lobe of the cerebellum. Increased microglia and astrocyte activation, as well as an accumulation of ubiquitinated proteins, p62 and TDP-43 were also detected in the cerebellum of aged TMEM106B deficient mice. In the young mice, myelination defects and a significant loss of synapses between Purkinje and deep cerebellar nuclei neurons were observed. Interestingly, TMEM106B deficiency causes distinct lysosomal phenotypes in different types of neurons and glia in the cerebellum and frontal cortex. In humans, TMEM106B rs1990622 risk allele (T/T) is associated with increased Purkinje neuron loss. Taken together, our studies support that TMEM106B regulates lysosomal function in a cell-type-specific manner and TMEM106B is critical for maintaining synaptic integrity and neural functions in the cerebellum.
Sigma-Aldrich
Anti-Parvalbumin antibody, Mouse monoclonal, clone PARV-19, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Ankyrin-G Antibody, clone N106/36, clone N106/36, from mouse
Sigma-Aldrich
Anti-Myelin Proteolipid Protein Antibody, CT, clone PLPC1, clone PLPC1, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse