Merck
  • HIV influences microtubule associated protein-2: potential marker of HIV-associated neurocognitive disorders.
HIV influences microtubule associated protein-2: potential marker of HIV-associated neurocognitive disorders.
AIDS (London, England) (2020-02-20)
Valeria Avdoshina, Matthew Mahoney, Sean F Gilmore, Erin D Wenzel, Albert Anderson, Scott L Letendre, Tomozumi Imamichi, Nicholas O Fischer, Italo Mocchetti
ABSTRACT
Postmortem brains of patients diagnosed with HIV-1-associated neurocognitive disorders (HAND) exhibit loss of dendrites. However, the mechanisms by which synapses are damaged are not fully understood. Dendrite length and remodeling occurs via microtubules, the dynamics of which are regulated by microtubule-binding proteins, including microtubule-associated protein 2 (MAP2). The HIV protein gp120 is neurotoxic and interferes with neuronal microtubules. We measured MAP2 concentrations in human cerebrospinal fluid (CSF) and MAP2 immunoreactivity in rat cortical neurons exposed to HIV and gp120. First, we examined whether HIV affects MAP2 levels by analyzing the CSF of 27 persons living with HIV (PLH) whose neurocognitive performance had been characterized. We then used rat cortical neurons to study the mechanisms of HIV-mediated dendritic loss. PLH who had HAND had greater MAP2 concentrations within the CSF than cognitive normal PLH. In cortical neurons, the deleterious effect of HIV on MAP2-positive dendrites occurred through a gp120-mediated mechanism. The neurotoxic effect of HIV was blocked by a CCR5 antagonist and prevented by Helix-A, a peptide that displaces gp120 from binding to microtubules, conjugated to a nanolipoprotein particle delivery platform. Our findings support that HIV at least partially effects its neurotoxicity via neuronal cytoskeleton modifications and provide evidence of a new therapeutic compound that could be used to prevent the HIV-associated neuropathology.
Sigma-Aldrich
DAPTA trifluoroacetate, ≥95% (HPLC)