Merck
  • Fluorescent detection of peroxynitrite during antibody-dependent cellular phagocytosis.
Fluorescent detection of peroxynitrite during antibody-dependent cellular phagocytosis.
Methods in enzymology (2020-06-21)
Digamber Rane, Erick J Carlson, Yuwen Yin, Blake R Peterson
ABSTRACT
Peroxynitrite (PNT) is a highly reactive oxidant that plays a key role in the destruction of foreign pathogens by specific phagocytic immune cells such as macrophages. However, when its production is dysregulated, this oxidant can contribute to cardiovascular disease, neurological diseases, and cancer. To facilitate the detection of PNT in living cells, we designed and synthesized a fluorescent sensor termed PS3 that accumulates in membranes of the endoplasmic reticulum (ER). This subcellular targeting enhances the proximity of PS3 to the phagosome of macrophages where PNT is generated. When PS3-treated macrophages are stimulated with 10 µm opsonized tentagel microspheres, antibody-dependent cellular phagocytosis (ADCP) of these particles results in production of endogenous PNT, oxidative cleavage of the fluorescence-quenching phenolic side chain of PS3, and increased fluorescence that can be detected by confocal laser scanning microscopy, flow cytometry, and other assays. We describe methods for the synthesis of PS3 and evaluation of its photophysical properties, selectivity, and reactivity. We further report differential production of PNT during ADCP by the phagocytic cell lines RAW 264.7, J774A.1, and THP-1, as detected by confocal microscopy and changes in fluorescence intensity on 96-well plates. This approach may be useful for identification of modulators of PNT and related studies of ADCP.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology