Merck
  • IFN-gamma induces high mobility group box 1 protein release partly through a TNF-dependent mechanism.
IFN-gamma induces high mobility group box 1 protein release partly through a TNF-dependent mechanism.
Journal of immunology (Baltimore, Md. : 1950) (2003-03-21)
Beatriz Rendon-Mitchell, Mahendar Ochani, Jianhua Li, Jialian Han, Hong Wang, Huan Yang, Seenu Susarla, Christopher Czura, Robert A Mitchell, Guoqian Chen, Andrew E Sama, Kevin J Tracey, Haichao Wang
ABSTRACT
We recently discovered that a ubiquitous protein, high mobility group box 1 protein (HMGB1), is released by activated macrophages, and functions as a late mediator of lethal systemic inflammation. To elucidate mechanisms underlying the regulation of HMGB1 release, we examined the roles of other cytokines in induction of HMGB1 release in macrophage cell cultures. Macrophage migration inhibitory factor, macrophage-inflammatory protein 1beta, and IL-6 each failed to significantly induce the release of HMGB1 even at supraphysiological levels (up to 200 ng/ml). IFN-gamma, an immunoregulatory cytokine known to mediate the innate immune response, dose-dependently induced the release of HMGB1, TNF, and NO, but not other cytokines such as IL-1alpha, IL-1beta, or IL-6. Pharmacological suppression of TNF activity with neutralizing Abs, or genetic disruption of TNF expression (TNF knockout) partially (50-60%) inhibited IFN-gamma-mediated HMGB1 release. AG490, a specific inhibitor for Janus kinase 2 of the IFN-gamma signaling pathway, dose-dependently attenuated IFN-gamma-induced HMGB1 release. These data suggest that IFN-gamma plays an important role in the regulation of HMGB1 release through a TNF- and Janus kinase 2-dependent mechanism.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
AG 490, A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC₅₀ = 100 nM).