Merck
  • Ibuprofen-Loaded Heparin Modified Thermosensitive Hydrogel for Inhibiting Excessive Inflammation and Promoting Wound Healing.
Ibuprofen-Loaded Heparin Modified Thermosensitive Hydrogel for Inhibiting Excessive Inflammation and Promoting Wound Healing.
Polymers (2020-11-12)
Abegaz Tizazu Andrgie, Haile Fentahun Darge, Tefera Worku Mekonnen, Yihenew Simegniew Birhan, Endiries Yibru Hanurry, Hsiao-Ying Chou, Chih-Feng Wang, Hsieh-Chih Tsai, Jen Ming Yang, Yen-Hsiang Chang
ABSTRACT
Hydrogels have been investigated as ideal biomaterials for wound treatment owing to their ability to form a highly moist environment which accelerates cell migration and tissue regeneration for prompt wound healing. They can also be used as a drug carrier for local delivery, and are able to activate immune cells to enhance wound healing. Here, we developed heparin-conjugated poly(N-isopropylacrylamide), an injectable, in situ gel-forming polymer, and evaluated its use in wound healing. Ibuprofen was encapsulated into the hydrogel to help reduce pain and excessive inflammation during healing. In addition to in vitro studies, a BALB/c mice model was used to evaluate its effect on would healing and the secretion of inflammatory mediators. The in vitro assay confirmed that the ibuprofen released from the hydrogel dramatically reduced lipopolysaccharide-induced inflammation by suppressing the production of NO, PGE2 and TNF-α in RAW264.7 macrophages. Moreover, an in vivo wound healing assay was conducted by applying hydrogels to wounds on the backs of mice. The results showed that the ibuprofen-loaded hydrogel improved healing relative to the phosphate buffered saline group. This study indicates that ibuprofen loaded in an injectable hydrogel is a promising candidate for wound healing therapy.
Product Description
Supelco
Ibuprofen sodium salt, ≥98% (GC)