Merck
  • Urinary monohydroxylated polycyclic aromatic hydrocarbons in primiparas from Shenzhen, South China: Levels, risk factors, and oxidative stress.
Urinary monohydroxylated polycyclic aromatic hydrocarbons in primiparas from Shenzhen, South China: Levels, risk factors, and oxidative stress.
Environmental pollution (Barking, Essex : 1987) (2020-01-10)
Mengmeng Peng, Shaoyou Lu, Yingxin Yu, Shan Liu, Yang Zhao, Chun Li, Shengtao Ma
ABSTRACT
The main objectives of the present study were to investigate urinary monohydroxylated polycyclic aromatic hydrocarbons (OH-PAHs) in 77 primiparas who live in Shenzhen, Guangdong Province, China, and their association with 8-hydroxy-2'-deoxyguanosine (8-OHdG) and human health risks. High detection frequencies of OH-PAHs demonstrated the wide occurrence of chemicals in the human exposure to PAHs. The urinary concentrations of Σ7OH-PAHs ranged from 1.37 to 45.5 ng/mL, and the median concentrations of 1-hydroxynaphthalene (1-OHN), 2-hydroxynaphthalene (2-OHN), 2-hydoxyfluorene (2-OHF), ΣOHPhe (the sum of 1-, 2+ 3-hydroxyphenanthrene), and 1-hydroxypyrene (1-OHP) were 3.00, 2.58, 0.31, 0.44, and 0.51 ng/mL, respectively. In the sum concentration of seven OH-PAHs, 1-OHN accounted for the largest proportion (43.7% of Σ7OH-PAHs), followed by 2-OHN (37.1%), 2-OHF (4.94%), 1-OHP (8.01%), 1-OHPhe (4.79%), and 2+3-OHPhe (1.46%). The present results showed that vehicle exhaust and petrochemical emission are the main sources of PAHs in primiparas in Shenzhen, and inhalation is the most important exposure route. The living conditions have a significant influence on human exposure to PAHs. The concentrations of 8-OHdG were positively correlated with OH-PAH concentrations in urine because evidence suggested that urinary 8-OHdG levels can be considered as a biomarker of oxidative DNA damage. Hazard quotient was used to assess the human health risks from exposure to single compound, and hazard index was used to assess the cumulative risks of the compounds, which demonstrated that the exposure risks from PAHs in primiparas were relatively low.
Sigma-Aldrich
Vinyl pivalate, contains hydroquinone as inhibitor, 99%
Sigma-Aldrich
2-Hydroxyfluorene, 98%