Merck
Toward fluorescence nanoscopy.
Nature biotechnology (2003-11-05)
Stefan W Hell
ABSTRACT
For more than a century, the resolution of focusing light microscopy has been limited by diffraction to 180 nm in the focal plane and to 500 nm along the optic axis. Recently, microscopes have been reported that provide three- to sevenfold improved axial resolution in live cells. Moreover, a family of concepts has emerged that overcomes the diffraction barrier altogether. Its first exponent, stimulated emission depletion microscopy, has so far displayed a resolution down to 28 nm. Relying on saturated optical transitions, these concepts are limited only by the attainable saturation level. As strong saturation should be feasible at low light intensities, nanoscale imaging with focused light may be closer than ever.
Sigma-Aldrich
Abberior® FLIP 565, maleimide, for single-molecule switching microscopy (e.g. PALM, STORM, GSDIM)
Sigma-Aldrich
Abberior® STAR 635, NHS ester, for STED application
Sigma-Aldrich
Abberior® STAR 580, NHS ester, for STED application
Sigma-Aldrich
Abberior® STAR 512, maleimide, for STED application
Sigma-Aldrich
Abberior® STAR 440SXP, maleimide, for long Stokes STED and 2-color STED application
Sigma-Aldrich
Abberior® STAR 440SXP, NHS ester, for long Stokes STED and 2-color STED application