Merck
  • The Effects of Metformin on Age-Related Changes in the Liver Sinusoidal Endothelial Cell.
The Effects of Metformin on Age-Related Changes in the Liver Sinusoidal Endothelial Cell.
The journals of gerontology. Series A, Biological sciences and medical sciences (2019-06-15)
Nicholas J Hunt, Glen P Lockwood, Sun Woo Sophie Kang, Tamara Pulpitel, Ximonie Clark, Hong Mao, Peter A G McCourt, Gregory J Cooney, Jibran A Wali, Frank H Le Couteur, David G Le Couteur, Victoria C Cogger
ABSTRACT
Age-related changes in the liver sinusoidal endothelium, particularly the reduction in fenestrations, contribute to insulin resistance in old age. Metformin impacts on the aging process and improves insulin resistance. Therefore, the effects of metformin on the liver sinusoidal endothelium were studied. Metformin increased fenestrations in liver sinusoidal endothelial cells isolated from both young and old mice. Mice administered metformin in the diet for 12 months had increased fenestrations and this was associated with lower insulin levels. The effect of metformin on fenestrations was blocked by inhibitors of AMP-activated protein kinase (AMPK), endothelial nitric oxide synthase, and myosin light chain kinase phosphorylation. Metformin led to increased transgelin expression and structural changes in the actin cytoskeleton but had no effect on lactate production. Metformin also generated fenestration-like structures in SK-Hep1 cells, a liver endothelial cell line, and this was associated with increased ATP, cGMP, and mitochondrial activity. In conclusion, metformin ameliorates age-related changes in the liver sinusoidal endothelial cell via AMPK and endothelial nitric oxide pathways, which might promote insulin sensitivity in the liver, particularly in old age.
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, with ribonucleosides, deoxyribonucleosides and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Myosin Light Chain Kinase Inhibitor Peptide 18, The Myosin Light Chain Kinase Inhibitor Peptide 18 controls the biological activity of Myosin Light Chain Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.