Merck
  • Increase of circulating IGFBP-4 following genotoxic stress and its implication for senescence.
Increase of circulating IGFBP-4 following genotoxic stress and its implication for senescence.
eLife (2020-04-01)
Nicola Alessio, Tiziana Squillaro, Giovanni Di Bernardo, Giovanni Galano, Roberto De Rosa, Mariarosa Ab Melone, Gianfranco Peluso, Umberto Galderisi
ABSTRACT
Senescent cells secrete several molecules, collectively named senescence-associated secretory phenotype (SASP). In the SASP of cells that became senescent following several in vitro chemical and physical stress, we identified the IGFBP-4 protein that can be considered a general stress mediator. This factor appeared to play a key role in senescence-paracrine signaling. We provided evidences showing that genotoxic injury, such as low dose irradiation, may promote an IGFBP-4 release in bloodstream both in mice irradiated with 100 mGy X-ray and in human subjects that received Computer Tomography. Increased level of circulating IGFBP-4 may be responsible of pro-aging effect. We found a significant increase of senescent cells in the lungs, heart, and kidneys of mice that were intraperitoneally injected with IGFBP-4 twice a week for two months. We then analyzed how genotoxic stressors may promote the release of IGFBP-4 and the molecular pathways associated with the induction of senescence by this protein.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-RB1 antibody produced in rabbit, affinity isolated antibody