Merck
  • Reduced plasma desmosterol-to-cholesterol ratio and longitudinal cognitive decline in Alzheimer's disease.
Reduced plasma desmosterol-to-cholesterol ratio and longitudinal cognitive decline in Alzheimer's disease.
Alzheimer's & dementia (Amsterdam, Netherlands) (2015-03-01)
Yoshiaki Sato, Francois Bernier, Yasukazu Yamanaka, Ken Aoshima, Yoshiya Oda, Martin Ingelsson, Lars Lannfelt, Akinori Miyashita, Ryozo Kuwano, Takeshi Ikeuchi
ABSTRACT
We here examined whether plasma desmosterol-to-cholesterol ratio (DES/CHO) is decreased in patients with Alzheimer's disease (AD) and investigated the association between plasma DES/CHO and longitudinal cognitive decline. Plasma DES/CHO of AD patients and age-matched controls in a Japanese cross-sectional cohort was determined. Plasma DES/CHO at baseline and follow-up visits was assessed in relation to cognitive decline in Japanese and Swedish longitudinal cohorts. Plasma DES/CHO was significantly reduced in Japanese AD patients and significantly correlated with Mini-Mental State Examination (MMSE) score. The longitudinal analysis revealed that plasma DES/CHO in AD patients shows a significant decrease at follow-up intervals. The decline in plasma DES/CHO is larger in the AD group with rapid progression than in that with slow progression. The changes in plasma DES/CHO significantly correlated with changes in the MMSE score. Plasma DES/CHO is decreased in AD patients and may serve as a longitudinal surrogate marker associated with cognitive decline.
Product Description
Avanti
desmosterol-d6, Avanti Research - A Croda Brand