Merck
  • Generation of induced Pluripotent Stem Cells (UNIBSi008-A, UNIBSi008-B, UNIBSi008-C) from an Ataxia-Telangiectasia (AT) patient carrying a novel homozygous deletion in ATM gene.
Generation of induced Pluripotent Stem Cells (UNIBSi008-A, UNIBSi008-B, UNIBSi008-C) from an Ataxia-Telangiectasia (AT) patient carrying a novel homozygous deletion in ATM gene.
Stem cell research (2019-11-02)
S Masneri, R M Ferraro, G Lanzi, G Piovani, L Mori, C Barisani, D Moratto, A Plebani, R Badolato, A Soresina, S Giliani
ABSTRACT
Using a Sendai Virus based vector delivering Yamanaka Factors, we generated induced Pluripotent Stem Cells (iPSCs) from peripheral blood mononuclear cells of a patient affected by Ataxia Telangiectasia (AT), caused by a novel homozygous deletion in ATM, spanning exons 5-7. Three clones were fully characterized for pluripotency and capability to differentiate. These clones preserved the causative mutation of parental cells and genomic stability over time (>100 passages). Furthermore, in AT derived iPSCs we confirmed the impaired DNA damage response after ionizing radiation. All these data underline potential usefulness of our clones as in vitro AT disease model.
Sigma-Aldrich
Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12 PE conjugate, clone 10H11.E12, from mouse, PE
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, FITC conjugate, clone JBW301, UpstateĀ®, from mouse