Merck
  • Genetically reducing mTOR signaling rescues central insulin dysregulation in a mouse model of Alzheimer's disease.
Genetically reducing mTOR signaling rescues central insulin dysregulation in a mouse model of Alzheimer's disease.
Neurobiology of aging (2018-05-08)
Antonella Caccamo, Ramona Belfiore, Salvatore Oddo
ABSTRACT
Alzheimer's disease (AD) is the most common neurodegenerative disease. The causes of sporadic AD, which represents more than 95% of AD cases, are unknown. Several AD risk factors have been identified and among these, type 2 diabetes increases the risk of developing AD by 2-fold. However, the mechanisms by which diabetes contributes to AD pathogenesis remain elusive. The mammalian target of rapamycin (mTOR) is a protein kinase that plays a crucial role in the insulin signaling pathway and has been linked to AD. We used a crossbreeding strategy to remove 1 copy of the mTOR gene from the forebrain of Tg2576 mice, a mouse model of AD. We used 20-month-old mice to assess changes in central insulin signaling and found that Tg2576 mice had impaired insulin signaling. These impairments were mTOR dependent as we found an improvement in central insulin signaling in mice with lower brain mTOR activity. Furthermore, removing 1 copy of mTOR from Tg2576 mice improved cognition and reduced levels of Aβ, tau, and cytokines. Our findings indicate that mTOR signaling is a key mediator of central insulin dysfunction in Tg2576. These data further highlight a possible role for mTOR signaling in AD pathogenesis and add to the body of evidence indicating that reducing mTOR activity could be a valid therapeutic approach for AD.
Sigma-Aldrich
Anti-Tau Mouse mAb (TAU-5), liquid, clone TAU-5, Calbiochem®
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®