Merck
  • Inexpensive but Highly Efficient Co-Mn Mixed-Oxide Catalysts for Selective Oxidation of 5-Hydroxymethylfurfural to 2,5-Furandicarboxylic Acid.
Inexpensive but Highly Efficient Co-Mn Mixed-Oxide Catalysts for Selective Oxidation of 5-Hydroxymethylfurfural to 2,5-Furandicarboxylic Acid.
ChemSusChem (2018-07-15)
Kasanneni Tirumala Venkateswara Rao, Jennifer Lorraine Rogers, Sadra Souzanchi, Luana Dessbesell, Madhumita Bhowmick Ray, Chunbao Charles Xu
ABSTRACT
A highly active and inexpensive Co-Mn mixed-oxide catalyst was prepared and used for selective oxidation of 5-hydroxymethylfurfural (HMF) into 2, 5-furandicarboxylic acid (FDCA). Co-Mn mixed-oxide catalysts with different Co/Mn molar ratios were prepared through a simple solid-state grinding method-a low-cost and green catalyst preparation method. The activity of these catalysts was evaluated for selective aerobic oxidation of HMF into FDCA in water. Excellent HMF conversion (99 %) and FDCA yield (95 % ) were obtained under the best reaction conditions (i.e., 120 °C, 5 h, Co-Mn mixed-oxide catalyst with a Co/Mn molar ratio of 0.25 calcined at 300 °C (Co-Mn-0.25) and 1 MPa O2 ). The catalyst could be reused five times without a significant decrease in activity. The results demonstrated that the catalytic activity and selectivity of the Co-Mn mixed-oxide catalysts prepared through solid-state grinding were superior to the same Co-Mn catalyst prepared through a conventional coprecipitation method. The high catalytic activity of the Co-Mn-0.25 catalyst was attributed to its high lattice oxygen mobility and the presence of different valence states of manganese. The high activity and low cost of the Co-Mn mixed-oxide catalysts prepared by solid-state grinding make it promising for industrial application for the manufacturing of polyethylene furanoate, a bioreplacement for polyethylene terephthalate, from sustainable bioresources.
Sigma-Aldrich
2,5-Furandicarboxylic acid, 97%
Sigma-Aldrich
2,5-Furandicarboxaldehyde, 97%