Merck
  • ASTN2 modulates synaptic strength by trafficking and degradation of surface proteins.
ASTN2 modulates synaptic strength by trafficking and degradation of surface proteins.
Proceedings of the National Academy of Sciences of the United States of America (2018-09-23)
Hourinaz Behesti, Taylor R Fore, Peter Wu, Zachi Horn, Mary Leppert, Court Hull, Mary E Hatten
ABSTRACT
Surface protein dynamics dictate synaptic connectivity and function in neuronal circuits. ASTN2, a gene disrupted by copy number variations (CNVs) in neurodevelopmental disorders, including autism spectrum, was previously shown to regulate the surface expression of ASTN1 in glial-guided neuronal migration. Here, we demonstrate that ASTN2 binds to and regulates the surface expression of multiple synaptic proteins in postmigratory neurons by endocytosis, resulting in modulation of synaptic activity. In cerebellar Purkinje cells (PCs), by immunogold electron microscopy, ASTN2 localizes primarily to endocytic and autophagocytic vesicles in the cell soma and in subsets of dendritic spines. Overexpression of ASTN2 in PCs, but not of ASTN2 lacking the FNIII domain, recurrently disrupted by CNVs in patients, including in a family presented here, increases inhibitory and excitatory postsynaptic activity and reduces levels of ASTN2 binding partners. Our data suggest a fundamental role for ASTN2 in dynamic regulation of surface proteins by endocytic trafficking and protein degradation.
Sigma-Aldrich
Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt, ≥95% (H-NMR), powder
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Gold Chloride Solution, 200 mg/dL in deionized water
Sigma-Aldrich
Suberic acid bis(N-hydroxysuccinimide ester), ≥95%, powder