Merck
  • Large-scale identification and visualization of N-glycans with primary structures using GlySeeker.
Large-scale identification and visualization of N-glycans with primary structures using GlySeeker.
Rapid communications in mass spectrometry : RCM (2017-11-07)
Kaijie Xiao, Yue Wang, Yun Shen, Yuyin Han, Zhixin Tian
ABSTRACT
Most of the current popular tandem mass spectrometers have the capability of resolving the primary structures (monosaccharide composition, sequence and linkage) of N-glycans; however, compositions or putative structures have mostly been reported so far. Identification and visualization tools of N-glycans are needed. The isotopic mass-to-charge ratio and envelope fingerprinting algorithm, which has been successfully used for intact protein database search and identification, was adapted for N-glycan database search, and a stand-alone N-glycan database search engine, GlySeeker, for automated N-glycan identification and visualization was developed and successfully benchmarked. Both pseudo 2D graph and one-line text formats with one-letter symbols for monosaccharides were proposed for representing N-glycans. N-glycans were identified with comprehensive interpretation of product ions and false discovery rate (FDR) control. In a database search of reversed-phase liquid chromatography/tandem mass spectrometry (RPLC/MS/MS) datasets of the N-glycome enriched from OVCAR-3 ovarian cancer cells, with FDR ≤1% and number of best hits (NoBHs) = 1-30, 1525 N-glycans with comprehensive primary structural information (composition, sequence and linkage) were identified and visualized; among these 1525 N-glycans, 559 had NoBHs = 1, i.e. their structures were uniquely identified. This represents a large-scale identification and visualization of N-glycans with primary structures from tandem mass spectra. A stand-alone N-glycan database search engine called GlySeeker has been developed for large-scale identification and visualization of N-glycans with comprehensive interpretation of tandem mass spectra and FDR control.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16