
Tropolone

Synonym(s):
2-Hydroxy-2,4,6-cycloheptatrien-1-one
Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Showing 1-3 of 3